Product Details

SNP ID
rs12361429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111519722 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCTATTTAAGAGAATCTTCTACA[C/T]GTTTTGAACATATACATTGATAAGT
Phenotype
MIM: 605673 MIM: 611374 MIM: 611375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTG4 PubMed Links

Gene Details

Gene
BTG4
Gene Name
BTG anti-proliferation factor 4
There are no transcripts associated with this gene.

Gene
C11orf88
Gene Name
chromosome 11 open reading frame 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100388.1 Intron NP_001093858.1
NM_207430.2 Intron NP_997313.2
Gene
MIR34B
Gene Name
microRNA 34b
There are no transcripts associated with this gene.

Gene
MIR34C
Gene Name
microRNA 34c
There are no transcripts associated with this gene.

View Full Product Details