Product Details

SNP ID
rs12808742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111603996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGAAGGGGTATATGTCATTCTG[G/T]CTTCTTTACTTTTTGCCTTCCCCTG
Phenotype
MIM: 608973
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIK2 PubMed Links

Gene Details

Gene
SIK2
Gene Name
salt inducible kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015191.2 Intron NP_056006.1
XM_017017417.1 Intron XP_016872906.1
XM_017017418.1 Intron XP_016872907.1

View Full Product Details