Product Details

SNP ID
rs11218950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123072200 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGCAATCCCTGATCTTTTTGATA[A/G]TTTAAGGCTTAGAATCATCAAAAGT
Phenotype
MIM: 611693 MIM: 600816
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLMP PubMed Links

Gene Details

Gene
CLMP
Gene Name
CXADR like membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024769.3 2761 UTR 3 NP_079045.1
XM_011542998.2 2761 Intron XP_011541300.1
Gene
HSPA8
Gene Name
heat shock protein family A (Hsp70) member 8
There are no transcripts associated with this gene.

View Full Product Details