Product Details
- SNP ID
-
rs11827843
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:124038964 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCCTGGGGCATGGGGAAGGCCC[A/G]TGAGGATGAACGCTGTCAGTAGGGT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10G7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11219420] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10G7
- Gene Name
- olfactory receptor family 10 subfamily G member 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004463.1 |
38 |
Missense Mutation |
ACG,ATG |
T13M |
NP_001004463.1 |
- Gene
- OR10G8
- Gene Name
- olfactory receptor family 10 subfamily G member 8
There are no transcripts associated with this gene.
View Full Product Details