Product Details

SNP ID
rs11031336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31265841 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGAAAGAAGATTCAACAAATA[C/T]TAAATACTGGTTATCTATTATTTAA
Phenotype
MIM: 608062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCDC1 PubMed Links

Gene Details

Gene
DCDC1
Gene Name
doublecortin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181807.3 Intron NP_861523.2

View Full Product Details