Product Details

SNP ID
rs10835981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32834453 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTTAGAATACTTTTTAAAACAA[C/T]TATCACCAATTATCCCATTGTAGCT
Phenotype
MIM: 611690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRRG4 PubMed Links

Gene Details

Gene
PRRG4
Gene Name
proline rich and Gla domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024081.5 Intron NP_076986.1
XM_006718313.3 Intron XP_006718376.3
XM_006718314.3 Intron XP_006718377.1
XM_017018275.1 Intron XP_016873764.1

View Full Product Details