Product Details
- SNP ID
-
rs10840228
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:9386879 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGGCCTTTATTTTTGTAATCCTTA[A/G]CTATTTACATTTCATACATTTCTTT
- Phenotype
-
MIM: 605586
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IPO7
PubMed Links
Gene Details
- Gene
- IPO7
- Gene Name
- importin 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006391.2 |
|
Intron |
|
|
NP_006382.1 |
View Full Product Details