Product Details

SNP ID
rs11575008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8052985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTCCTCAGTCCTTACCACTTC[C/T]CTCCACGACCCCAGGTGGCCCCGTT
Phenotype
MIM: 602565
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL25 PubMed Links

Gene Details

Gene
CCL25
Gene Name
C-C motif chemokine ligand 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201359.1 Intron NP_001188288.1
NM_005624.3 Intron NP_005615.2
XM_011528177.2 Intron XP_011526479.1
XM_017027119.1 Intron XP_016882608.1
XM_017027120.1 Intron XP_016882609.1
XM_017027121.1 Intron XP_016882610.1
XM_017027122.1 Intron XP_016882611.1

View Full Product Details