Product Details

SNP ID
rs12983762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4682987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCGGCAAGGAAGGGGCCCTCA[A/G]ACCGCGTGGCCCCCGTGGACGGTGC
Phenotype
MIM: 608258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPP9 PubMed Links

Gene Details

Gene
DPP9
Gene Name
dipeptidyl peptidase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139159.4 445 Intron NP_631898.3
XM_005259673.2 445 Intron XP_005259730.1
XM_011528405.1 445 Intron XP_011526707.1
XM_011528406.1 445 Intron XP_011526708.1
XM_011528408.1 445 Intron XP_011526710.1
XM_011528410.1 445 Intron XP_011526712.1
XM_017027453.1 445 Intron XP_016882942.1
XM_017027454.1 445 Intron XP_016882943.1
Gene
DPP9-AS1
Gene Name
DPP9 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242901.1 445 Silent Mutation CAA,CAG Q43Q NP_001229830.1

View Full Product Details