Product Details

SNP ID
rs12461200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4677205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAACTCCCCAGGCTCCTGAGAC[C/T]TGGACCAGCCAACGCCAGGCCTGGG
Phenotype
MIM: 608258 MIM: 606746
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPP9 PubMed Links

Gene Details

Gene
DPP9
Gene Name
dipeptidyl peptidase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139159.4 Intron NP_631898.3
XM_005259673.2 Intron XP_005259730.1
XM_011528405.1 Intron XP_011526707.1
XM_011528406.1 Intron XP_011526708.1
XM_011528408.1 Intron XP_011526710.1
XM_011528410.1 Intron XP_011526712.1
XM_017027453.1 Intron XP_016882942.1
XM_017027454.1 Intron XP_016882943.1
Gene
DPP9-AS1
Gene Name
DPP9 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MYDGF
Gene Name
myeloid derived growth factor
There are no transcripts associated with this gene.

View Full Product Details