Product Details

SNP ID
rs12132981
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:236518885 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATCATGTTAACAAAAGTGACACA[A/G]CAACAAAACAATAAGCAAAATCACT
Phenotype
MIM: 606099
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGALS8 PubMed Links

Gene Details

Gene
LGALS8
Gene Name
galectin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006499.4 Intron NP_006490.3
NM_201543.2 Intron NP_963837.1
NM_201544.2 Intron NP_963838.1
NM_201545.2 Intron NP_963839.1
XM_011544188.2 Intron XP_011542490.1
XM_017001274.1 Intron XP_016856763.1
XM_017001275.1 Intron XP_016856764.1
Gene
LGALS8-AS1
Gene Name
LGALS8 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details