Product Details

SNP ID
rs6756580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202266218 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGATTTTATTTCCTCCCGTTGAAG[A/G]GGTGGGCAGGAGGCGTGTTAGAATT
Phenotype
MIM: 616742
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOP58 PubMed Links

Gene Details

Gene
NOP58
Gene Name
NOP58 ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015934.4 Intron NP_057018.1

View Full Product Details