Product Details
- SNP ID
-
rs13394555
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:237574679 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGACGTAAGGCATCTTCCCACCGT[C/T]GCTGTGCTGCGGGGACTTTTCCAAT
- Phenotype
-
MIM: 602663
MIM: 602206
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRLH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8184] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRLH
- Gene Name
- prolactin releasing hormone
There are no transcripts associated with this gene.
- Gene
- RAB17
- Gene Name
- RAB17, member RAS oncogene family
View Full Product Details