Product Details

SNP ID
rs9658047
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:7920057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGACCCTAAACTCAGGCACTCTG[C/T]TCATACTCCCCCCTGACGGTGGAGC
Phenotype
MIM: 602250 MIM: 604097
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF9 PubMed Links

Gene Details

Gene
TNFRSF9
Gene Name
TNF receptor superfamily member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001561.5 1807 UTR 3 NP_001552.2
XM_006710618.3 1807 Intron XP_006710681.1
XM_011541386.2 1807 UTR 3 XP_011539688.1
Gene
UTS2
Gene Name
urotensin 2
There are no transcripts associated with this gene.

View Full Product Details