Product Details

SNP ID
rs11582969
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:9306021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGGGAGAGGGGTTCGTGCTTTA[G/T]GGAACGAAGGCGTGTTGGGGACAGA
Phenotype
MIM: 611657
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPSB1 PubMed Links

Gene Details

Gene
SPSB1
Gene Name
splA/ryanodine receptor domain and SOCS box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025106.3 Intron NP_079382.2

View Full Product Details