Product Details

SNP ID
rs11227213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65526561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATTCGGGGTTGGGTGATTCTG[A/T]ACTTGAAAGCTCTGTGACCTGGACA
Phenotype
MIM: 607982
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SCYL1 PubMed Links

Gene Details

Gene
SCYL1
Gene Name
SCY1 like pseudokinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001048218.1 Intron NP_001041683.1
NM_020680.3 Intron NP_065731.3
XM_005274118.3 Intron XP_005274175.1
XM_005274120.3 Intron XP_005274177.1
XM_005274121.3 Intron XP_005274178.1
XM_017018029.1 Intron XP_016873518.1
XM_017018030.1 Intron XP_016873519.1
XM_017018031.1 Intron XP_016873520.1
XM_017018032.1 Intron XP_016873521.1
XM_017018033.1 Intron XP_016873522.1

View Full Product Details