Product Details
- SNP ID
-
rs7111253
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74752275 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGGGAAAAAAAAGAAGTGATTCT[C/G]TCTTACCCCCTTATTTCTCACTCCC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RNF169
PubMed Links
Gene Details
- Gene
- RNF169
- Gene Name
- ring finger protein 169
View Full Product Details