Product Details
- SNP ID
-
rs12290239
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74994257 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATGCAAAACCGCTCTGCTCCAAG[A/G]TACATACTTGATACAGGCCACATAG
- Phenotype
-
MIM: 604617
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NEU3
PubMed Links
Gene Details
- Gene
- NEU3
- Gene Name
- neuraminidase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006656.5 |
|
Intron |
|
|
NP_006647.3 |
View Full Product Details