Product Details

SNP ID
rs7265445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:34375880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAAATTAAAAAGGAAAGTTTTCA[A/G]AAGGTTAAAAAAAAAATTGAGGCAA
Phenotype
MIM: 606409
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITCH PubMed Links

Gene Details

Gene
ITCH
Gene Name
itchy E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257137.2 Intron NP_001244066.1
NM_001257138.2 Intron NP_001244067.1
NM_001324197.1 Intron NP_001311126.1
NM_001324198.1 Intron NP_001311127.1
NM_031483.6 Intron NP_113671.3
XM_005260570.1 Intron XP_005260627.1
XM_011529079.1 Intron XP_011527381.1
XM_017028089.1 Intron XP_016883578.1
XM_017028090.1 Intron XP_016883579.1
XM_017028091.1 Intron XP_016883580.1
XM_017028092.1 Intron XP_016883581.1
Gene
LOC107985381
Gene Name
uncharacterized LOC107985381
There are no transcripts associated with this gene.

View Full Product Details