Product Details

SNP ID
rs3814105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94657752 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATGAAATGAATCTGGCTTTGGTG[A/G]CTGGTGGGAGGGGTTTGGGAGGGGG
Phenotype
MIM: 609810 MIM: 604149
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PEG10 PubMed Links

Gene Details

Gene
PEG10
Gene Name
paternally expressed 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040152.1 Intron NP_001035242.1
NM_001172437.2 Intron NP_001165908.1
NM_001172438.2 Intron NP_001165909.1
NM_001184961.1 Intron NP_001171890.1
NM_001184962.1 Intron NP_001171891.1
NM_015068.3 Intron NP_055883.2
Gene
SGCE
Gene Name
sarcoglycan epsilon
There are no transcripts associated with this gene.

View Full Product Details