Product Details

SNP ID
rs11588791
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:244054417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGGAGAGGCAGAGCCACACGCC[A/T]CAGCAGCTGGAAAAACAGTAGCCAG
Phenotype
MIM: 608433
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC339529 PubMed Links

Gene Details

Gene
LOC339529
Gene Name
uncharacterized LOC339529
There are no transcripts associated with this gene.

Gene
LOC440742
Gene Name
uncharacterized LOC440742
There are no transcripts associated with this gene.

Gene
ZBTB18
Gene Name
zinc finger and BTB domain containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278196.1 746 Missense Mutation ACA,TCA T206S NP_001265125.1
NM_006352.4 746 Missense Mutation ACA,TCA T206S NP_006343.2
NM_205768.2 746 Missense Mutation ACA,TCA T215S NP_991331.1
XM_005273006.2 746 Missense Mutation ACA,TCA T206S XP_005273063.1
XM_017000060.1 746 Missense Mutation ACA,TCA T206S XP_016855549.1

View Full Product Details