Product Details

SNP ID
rs3131611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50507467 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTAGATATGACCAAGAGAAAGCT[C/G]AAGTAGCTATACATCAGCTAAAATG
Phenotype
MIM: 603158
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP50 PubMed Links

Gene Details

Gene
USP50
Gene Name
ubiquitin specific peptidase 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203494.4 Intron NP_987090.2
Gene
USP8
Gene Name
ubiquitin specific peptidase 8
There are no transcripts associated with this gene.

View Full Product Details