Product Details

SNP ID
rs12591532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55357437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATACCTTTCCCATCAATCCTCTA[A/G]GCTGCTACCAATACATTAGTCCCAC
Phenotype
MIM: 611326 MIM: 604122
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCPG1 PubMed Links

Gene Details

Gene
CCPG1
Gene Name
cell cycle progression 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204450.1 4875 Intron NP_001191379.1
NM_001204451.1 4875 Intron NP_001191380.1
NM_004748.4 4875 UTR 3 NP_004739.3
NM_020739.3 4875 UTR 3 NP_065790.2
Gene
DYX1C1-CCPG1
Gene Name
DYX1C1-CCPG1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
PIGB
Gene Name
phosphatidylinositol glycan anchor biosynthesis class B
There are no transcripts associated with this gene.

View Full Product Details