Product Details

SNP ID
rs12594039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55422328 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATCTGAGGAAAGTCAAGTTGATG[C/T]AAGGCAACTGAATATACAACTTGAA
Phenotype
MIM: 608706
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf65 PubMed Links

Gene Details

Gene
C15orf65
Gene Name
chromosome 15 open reading frame 65
There are no transcripts associated with this gene.

Gene
DYX1C1
Gene Name
dyslexia susceptibility 1 candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033559.2 Intron NP_001028731.1
NM_001033560.1 Intron NP_001028732.1
NM_130810.3 Intron NP_570722.2
Gene
DYX1C1-CCPG1
Gene Name
DYX1C1-CCPG1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details