Product Details

SNP ID
rs4633660
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:53515202 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTATTACAATGACAGGAAGACTC[C/T]GGATACAAACACATTTGCTAATATA
Phenotype
MIM: 613214
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
WDR72 PubMed Links

Gene Details

Gene
WDR72
Gene Name
WD repeat domain 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277176.1 6045 UTR 3 NP_001264105.1
NM_182758.3 6045 UTR 3 NP_877435.3
XM_011521433.2 6045 UTR 3 XP_011519735.1
XM_011521435.2 6045 UTR 3 XP_011519737.1
XM_011521436.2 6045 Intron XP_011519738.1
XM_011521437.2 6045 Intron XP_011519739.1
XM_017022061.1 6045 UTR 3 XP_016877550.1

View Full Product Details