Product Details
- SNP ID
-
rs12588179
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:76379795 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGGAGTTTGTTACTTCCCCCAAC[A/C]CTTTTCATTTTAGAAACAATCCTTT
- Phenotype
-
MIM: 602167
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ESRRB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111886499] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ESRRB
- Gene Name
- estrogen related receptor beta
View Full Product Details