Product Details

SNP ID
rs7160288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76961015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAACGCATTTCTTAGTGCAGGTC[A/T]TGATTTCAAATGTTCGAAAGCCACT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01629 PubMed Links

Gene Details

Gene
LINC01629
Gene Name
long intergenic non-protein coding RNA 1629
There are no transcripts associated with this gene.

View Full Product Details