Product Details

SNP ID
rs11626925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90541571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCGCCAGACTGTAGCGGCCTA[A/G]CTGGTGAAGGATCAGGGCCTGGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC7B PubMed Links

Gene Details

Gene
TTC7B
Gene Name
tetratricopeptide repeat domain 7B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010854.1 1975 Silent Mutation CTA,TTA L777L NP_001010854.1
NM_001320421.1 1975 Silent Mutation CTA,TTA L692L NP_001307350.1
XM_005267367.1 1975 Silent Mutation CTA,TTA L590L XP_005267424.1
XM_011536497.1 1975 Silent Mutation CTA,TTA L746L XP_011534799.1
XM_017021044.1 1975 Silent Mutation CTA,TTA L848L XP_016876533.1
XM_017021045.1 1975 Silent Mutation CTA,TTA L794L XP_016876534.1
XM_017021046.1 1975 Silent Mutation CTA,TTA L675L XP_016876535.1
XM_017021047.1 1975 Silent Mutation CTA,TTA L519L XP_016876536.1

View Full Product Details