Product Details

SNP ID
rs12433076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92324865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTGAGCAGTTAGAAAGGCCGTC[A/G]ACATCCAGAGACCATTTAGAGAGGT
Phenotype
MIM: 609840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC24A4 PubMed Links

Gene Details

Gene
SLC24A4
Gene Name
solute carrier family 24 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153646.3 Intron NP_705932.2
NM_153647.3 Intron NP_705933.2
NM_153648.3 Intron NP_705934.1
XM_005267342.2 Intron XP_005267399.1
XM_011536436.2 Intron XP_011534738.1
XM_011536437.2 Intron XP_011534739.1
XM_011536438.2 Intron XP_011534740.1
XM_011536439.2 Intron XP_011534741.1
XM_011536440.2 Intron XP_011534742.1
XM_011536441.2 Intron XP_011534743.1
XM_011536442.2 Intron XP_011534744.1

View Full Product Details