Product Details
- SNP ID
-
rs11667945
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:17441497 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGCAGAGTGGATTTGGGATCACTA[A/G]GCCACTTTGGCATCAATGATGTCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM221
PubMed Links
Gene Details
- Gene
- TMEM221
- Gene Name
- transmembrane protein 221
View Full Product Details