Product Details

SNP ID
rs12149723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3616725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTGTTCTTTTTCAAGAACGTA[C/T]TTCCTTATATCAAAACTTATTACAA
Phenotype
MIM: 125505 MIM: 613278
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNASE1 PubMed Links

Gene Details

Gene
DNASE1
Gene Name
deoxyribonuclease I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005223.3 Intron NP_005214.2
XM_005255148.3 Intron XP_005255205.1
XM_006720854.3 Intron XP_006720917.1
XM_011522393.2 Intron XP_011520695.1
XM_017022992.1 Intron XP_016878481.1
XM_017022993.1 Intron XP_016878482.1
XM_017022994.1 Intron XP_016878483.1
XM_017022995.1 Intron XP_016878484.1
XM_017022996.1 Intron XP_016878485.1
XM_017022997.1 Intron XP_016878486.1
XM_017022998.1 Intron XP_016878487.1
XM_017022999.1 Intron XP_016878488.1
XM_017023000.1 Intron XP_016878489.1
XM_017023001.1 Intron XP_016878490.1
XM_017023002.1 Intron XP_016878491.1
XM_017023003.1 Intron XP_016878492.1
XM_017023004.1 Intron XP_016878493.1
XM_017023005.1 Intron XP_016878494.1
XM_017023006.1 Intron XP_016878495.1
XM_017023007.1 Intron XP_016878496.1
Gene
SLX4
Gene Name
SLX4 structure-specific endonuclease subunit
There are no transcripts associated with this gene.

View Full Product Details