Product Details

SNP ID
rs4786460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1708578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGGCGCTGCCCAGCTCTGTGCGG[G/T]TCACATGCAAGCTGCTGTGAGTGCA
Phenotype
MIM: 605431
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HN1L PubMed Links

Gene Details

Gene
HN1L
Gene Name
hematological and neurological expressed 1-like
There are no transcripts associated with this gene.

Gene
MAPK8IP3
Gene Name
mitogen-activated protein kinase 8 interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040439.1 Intron NP_001035529.1
NM_001318852.1 Intron NP_001305781.1
NM_015133.4 Intron NP_055948.2

View Full Product Details