Product Details

SNP ID
rs11006814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:28053124 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGCAATTTAAAGAGGGGTGATCTA[C/T]TGCACATAGATGAAAAATAATCTTT
Phenotype
MIM: 610973
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPP7 PubMed Links

Gene Details

Gene
MPP7
Gene Name
membrane palmitoylated protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318170.1 2934 UTR 3 NP_001305099.1
NM_173496.4 2934 UTR 3 NP_775767.2
XM_005252369.3 2934 Intron XP_005252426.1
XM_011519337.2 2934 UTR 3 XP_011517639.1
XM_011519338.2 2934 UTR 3 XP_011517640.1
XM_017015741.1 2934 UTR 3 XP_016871230.1
XM_017015742.1 2934 UTR 3 XP_016871231.1
XM_017015743.1 2934 Intron XP_016871232.1

View Full Product Details