Product Details

SNP ID
rs10828708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:24852890 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACGACTGGTTCATGCTCATAGAAA[G/T]TACTGGAGGTACTTATTTTCTAGGG
Phenotype
MIM: 610751
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRTFDC1 PubMed Links

Gene Details

Gene
PRTFDC1
Gene Name
phosphoribosyl transferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282786.1 Intron NP_001269715.1
NM_020200.6 Intron NP_064585.1

View Full Product Details