Product Details

SNP ID
rs12767957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:18659653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTCGTGATGGGGCTGATCTTC[C/G]CCAAACTGTGGAGCCTCTTCTGTAA
Phenotype
MIM: 608909
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL5B PubMed Links

Gene Details

Gene
ARL5B
Gene Name
ADP ribosylation factor like GTPase 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178815.3 319 Missense Mutation CCC,GCC P6A NP_848930.1
XM_005252400.1 319 Missense Mutation CCC,GCC P6A XP_005252457.1
XM_005252401.4 319 Intron XP_005252458.1
Gene
NSUN6
Gene Name
NOP2/Sun RNA methyltransferase family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182543.3 319 Intron NP_872349.1
XM_011519383.1 319 Intron XP_011517685.1
XM_011519384.1 319 Intron XP_011517686.1
XM_011519385.1 319 Intron XP_011517687.1
XM_011519386.2 319 Intron XP_011517688.1
XM_011519387.2 319 Intron XP_011517689.1
XM_011519389.2 319 Intron XP_011517691.1
XM_017015906.1 319 Intron XP_016871395.1
XM_017015907.1 319 Intron XP_016871396.1
XM_017015908.1 319 Intron XP_016871397.1
XM_017015909.1 319 Intron XP_016871398.1
XM_017015910.1 319 Intron XP_016871399.1
XM_017015911.1 319 Intron XP_016871400.1
XM_017015912.1 319 Intron XP_016871401.1
XM_017015913.1 319 Intron XP_016871402.1
XM_017015914.1 319 Intron XP_016871403.1

View Full Product Details