Product Details

SNP ID
rs13377078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14880024 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAAAAGCACACAAACACACCTTG[A/G]GTAGATAAGAGGAACCTCCTTTATA
Phenotype
MIM: 610369 MIM: 606503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPA14 PubMed Links

Gene Details

Gene
HSPA14
Gene Name
heat shock protein family A (Hsp70) member 14
There are no transcripts associated with this gene.

Gene
SUV39H2
Gene Name
suppressor of variegation 3-9 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193424.1 295 Intron NP_001180353.1
NM_001193425.1 295 Intron NP_001180354.1
NM_001193426.1 295 Intron NP_001180355.1
NM_001193427.1 295 Intron NP_001180356.1
NM_024670.3 295 Intron NP_078946.1
XM_006717503.3 295 UTR 5 XP_006717566.1
XM_011519662.2 295 Intron XP_011517964.1
XM_017016637.1 295 Intron XP_016872126.1

View Full Product Details