Product Details

SNP ID
rs10906419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:13591835 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACTAGCCAGGTGCTTCAGGAAG[A/G]CCGCCCTCTTGCCTCTGTGGAATCC
Phenotype
MIM: 604993
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRPF18 PubMed Links

Gene Details

Gene
PRPF18
Gene Name
pre-mRNA processing factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003675.3 286 Intron NP_003666.1
XM_011519754.2 286 UTR 5 XP_011518056.1
XM_017016860.1 286 Intron XP_016872349.1

View Full Product Details