Product Details

SNP ID
rs12266469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:6345792 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTATCTTCTCTCTCTCTCTCAAT[C/T]GTCTGTTTATTGGTTGTTTCAAGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC399715 PubMed Links

Gene Details

Gene
LOC399715
Gene Name
uncharacterized LOC399715
There are no transcripts associated with this gene.

Gene
LOC399716
Gene Name
uncharacterized LOC399716
There are no transcripts associated with this gene.

View Full Product Details