Product Details

SNP ID
rs7076000
Assay Type
Functionally Tested
NCBI dbSNP Submissions
36
Location
Chr.10:1066429 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCGCCCAGCCCGATACTAGGACT[A/T]ATGCAGAAAAAACCTTGACATGGAG
Phenotype
MIM: 604055
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
IDI1 PubMed Links

Gene Details

Gene
IDI1
Gene Name
isopentenyl-diphosphate delta isomerase 1
There are no transcripts associated with this gene.

Gene
WDR37
Gene Name
WD repeat domain 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014023.3 Intron NP_054742.2

View Full Product Details