Product Details

SNP ID
rs8178252
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:47777997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTAAAAATACAGACTCTGCTTC[C/T]CTTGAAATCAGCTACACAGATGTGA
Phenotype
MIM: 600899
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PRKDC PubMed Links

Gene Details

Gene
PRKDC
Gene Name
protein kinase, DNA-activated, catalytic polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081640.1 Intron NP_001075109.1
NM_006904.6 Intron NP_008835.5

View Full Product Details