Product Details

SNP ID
rs12336832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113270528 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGCTATGAAAAAAATATGGTGC[C/T]TTGTGAGAGCCTACAGAGATGGAGG
Phenotype
MIM: 614533 MIM: 607795 MIM: 603085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC26 PubMed Links

Gene Details

Gene
CDC26
Gene Name
cell division cycle 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139286.3 Intron NP_644815.1
XM_017014573.1 Intron XP_016870062.1
XM_017014574.1 Intron XP_016870063.1
XM_017014575.1 Intron XP_016870064.1
Gene
PRPF4
Gene Name
pre-mRNA processing factor 4
There are no transcripts associated with this gene.

Gene
SLC31A1
Gene Name
solute carrier family 31 member 1
There are no transcripts associated with this gene.

View Full Product Details