Product Details

SNP ID
rs13302330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113151105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAGGGCCGGGCGCTACGGTGAA[A/G]AGGGTGGGCGGTTGGGGCGGGGTCT
Phenotype
MIM: 603088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC31A2 PubMed Links

Gene Details

Gene
SLC31A2
Gene Name
solute carrier family 31 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001860.2 Intron NP_001851.1
XM_011518216.1 Intron XP_011516518.1
XM_017014267.1 Intron XP_016869756.1

View Full Product Details