Product Details

SNP ID
rs12236936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113119384 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCACCACTCTCCACTTGGGAGGAG[G/T]CCTTAACATCCCCCTCAGGTCTAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM225A PubMed Links

Gene Details

Gene
FAM225A
Gene Name
family with sequence similarity 225 member A (non-protein coding)
There are no transcripts associated with this gene.

Gene
FAM225B
Gene Name
family with sequence similarity 225 member B (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details