Product Details
- SNP ID
-
rs12685234
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:114607315 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACTCTCTTTTTGGGAAAGGGGACA[C/T]TGAACAATGACAATACCATGTGATG
- Phenotype
-
MIM: 607296
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP6V1G1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116905709,rs78204542] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP6V1G1
- Gene Name
- ATPase H+ transporting V1 subunit G1
There are no transcripts associated with this gene.
- Gene
- TMEM268
- Gene Name
- transmembrane protein 268
View Full Product Details