Product Details

SNP ID
rs10985626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122372690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCATCTGGCCTCATCCAGGCAC[C/T]CTTGGTATACAGGTCCCACCCGTCT
Phenotype
MIM: 176805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTGS1 PubMed Links

Gene Details

Gene
PTGS1
Gene Name
prostaglandin-endoperoxide synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000962.3 1466 Intron NP_000953.2
NM_001271164.1 1466 Intron NP_001258093.1
NM_001271165.1 1466 Intron NP_001258094.1
NM_001271166.1 1466 Intron NP_001258095.1
NM_001271367.1 1466 Intron NP_001258296.1
NM_001271368.1 1466 Intron NP_001258297.1
NM_080591.2 1466 Intron NP_542158.1
XM_005252105.3 1466 Intron XP_005252162.1
XM_011518875.2 1466 Intron XP_011517177.1
XM_011518876.2 1466 UTR 5 XP_011517178.1
XM_017014929.1 1466 Intron XP_016870418.1

View Full Product Details