Product Details
- SNP ID
-
rs4988465
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:73169399 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGACCTCAATGGCTCTGTGGTGT[C/T]AACCAACACCTCAAACCAGACAGAG
- Phenotype
-
MIM: 182452
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SSTR2
PubMed Links
Gene Details
- Gene
- SSTR2
- Gene Name
- somatostatin receptor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001050.2 |
440 |
Missense Mutation |
TCA,TTA |
S27L |
NP_001041.1 |
View Full Product Details