Product Details

SNP ID
rs7210937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:12917329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAGAAGCCAGTAGCATGGCTCA[C/G]TCCAGAAACCTCAGAACCCGAAACT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP44 PubMed Links

Gene Details

Gene
ARHGAP44
Gene Name
Rho GTPase activating protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321164.1 Intron NP_001308093.1
NM_001321166.1 Intron NP_001308095.1
NM_001321167.1 Intron NP_001308096.1
NM_001321168.1 Intron NP_001308097.1
NM_014859.5 Intron NP_055674.4
XM_005256891.1 Intron XP_005256948.1
Gene
MIR1269B
Gene Name
microRNA 1269b
There are no transcripts associated with this gene.

View Full Product Details