Product Details
- SNP ID
-
hCV32137936
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:165405451 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCATACAACAGGATACATTCTCCA[G/T]CCTCCTTTGTGGCCAGCTGGGCCAC
- Phenotype
-
MIM: 180247
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RXRG
PubMed Links
Gene Details
- Gene
- RXRG
- Gene Name
- retinoid X receptor gamma
There are no transcripts associated with this gene.
View Full Product Details