Product Details

SNP ID
rs12042693
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:167544486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGTCTACCTTAGTTATCATCACC[A/G]TAATCTCCTGAAACAGCAGTCAATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CREG1 PubMed Links

Gene Details

Gene
CREG1
Gene Name
cellular repressor of E1A stimulated genes 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003851.2 Intron NP_003842.1

View Full Product Details