Product Details
- SNP ID
-
rs12077118
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:167734713 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAGAGAGGCGTTGGGGGTAGGTCC[C/T]GAGGAGAATCAGCAGTGTCTTGCAA
- Phenotype
-
MIM: 604376
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MPZL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79468622] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MPZL1
- Gene Name
- myelin protein zero like 1
View Full Product Details